6-Chloro-3-indolyl α-glucopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: Salmon α-glucoside

BIMB1329-100MG BIMB1329-250MG
APOSBIMB1329-100MGEA 102 EUR
APOSBIMB1329-100MG APOSBIMB1329-250MG
6-Chloro-3-indolyl α-glucopyranoside
6-Chloro-3-indolyl α-glucopyranoside
Formel: C₁₄H₁₆ClNO₆
Molecular Weight: 329,733 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD04972053
CAS-Nummer: 467214-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR