5-Bromo-6-chloro-3-indolyl caprylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Magenta caprylate

BIMB2110-100MG BIMB2110-1G BIMB2110-250MG
APOSBIMB2110-100MGEA 36.4 EUR
APOSBIMB2110-100MG APOSBIMB2110-1G APOSBIMB2110-250MG
5-Bromo-6-chloro-3-indolyl caprylate
5-Bromo-6-chloro-3-indolyl caprylate

Histochemical substrate for esterase

Formel: C₁₆H₁₉BrClNO₂
Molecular Weight: 372,685 g/mol
Schmelzpunkt: 57…62 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00798386
CAS-Nummer: 209347-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR