Propionylleupeptin hemisulphate

Lieferant: Apollo Scientific
Achtung

Synonyme: Propionyl-Leu-Leu-Arg-al hemisulphate

BIMI0874-100MG BIMI0874-25MG
APOSBIMI0874-100MGEA 232 EUR
APOSBIMI0874-100MG APOSBIMI0874-25MG
Propionylleupeptin hemisulphate
Propionylleupeptin hemisulphate

Inhibitor of serine and cysteine proteases

Lagertemperatur: Gefrierschrank
MDL: MFCD00171338
CAS-Nummer: 24365-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR