D-Glucose-6-phosphate trisodium salt
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Robison ester trisodium salt
BIP0687-50MG
APOSBIP0687-50MGEA
50.9
EUR
APOSBIP0687-50MG
D-Glucose-6-phosphate trisodium salt
D-Glucose-6-phosphate trisodium salt
Indefinitely stable at -15°C
Formel:
C₆H₁₀O₁₀P·3Na Molecular Weight: 342,081 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00083681 CAS-Nummer: 53411-70-4 EINECS: 258-543-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...