Amino-PEG9-acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Amino-3,6,9,12,15,18,21,24,27-nonaoxatriacontan-30-oic acid

BIPG1037-100MG BIPG1037-50MG BIPG1037-250MG
APOSBIPG1037-100MGEA 212 EUR
APOSBIPG1037-100MG APOSBIPG1037-50MG APOSBIPG1037-250MG
Amino-PEG9-acid
Amino-PEG9-acid
Formel: C₂₁H₄₃NO₁₁
Molecular Weight: 485.566 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD26127809
CAS-Nummer: 1191079-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR