Amino-PEG7-alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
NH2-PEG7, 20-Amino-3,6,9,12,15,18-hexaoxaicosan-1-ol
BIPG1049-100MG
BIPG1049-250MG
BIPG1049-500MG
APOSBIPG1049-100MGEA
87
EUR
APOSBIPG1049-100MG
APOSBIPG1049-250MG
APOSBIPG1049-500MG
Amino-PEG7-alcohol
Amino-PEG7-alcohol
Formel:
C₁₄H₃₁NO₇ Molecular Weight: 325.398 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28385479 CAS-Nummer: 1425973-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...