Amino-PEG4-CH2CO2tBu

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 14-amino-3,6,9,12-tetraoxatetradecanoate, Amino-PEG4-C1-Boc

BIPG1068-100MG BIPG1068-250MG BIPG1068-500MG
APOSBIPG1068-100MGEA 109 EUR
APOSBIPG1068-100MG APOSBIPG1068-250MG APOSBIPG1068-500MG
Amino-PEG4-CH2CO2tBu
Amino-PEG4-CH2CO2tBu
Formel: C₁₄H₂₉NO₆
Molecular Weight: 307.383 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28122943
CAS-Nummer: 864680-64-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR