Azido-PEG6-Alcohol
Lieferant: Apollo Scientific
|
Achtung
|
BIPG1119-100MG
BIPG1119-50MG
BIPG1119-250MG
APOSBIPG1119-100MGEA
148
EUR
APOSBIPG1119-100MG
APOSBIPG1119-50MG
APOSBIPG1119-250MG
Azido-PEG6-Alcohol
Azido-PEG6-Alcohol
Formel:
C₁₂H₂₅N₃O₆ Molecular Weight: 307.343 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22056309 CAS-Nummer: 86770-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...