Biotin-PEG3-acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 14-Oxo-18-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10-trioxa-13-azaoctadecanoic acid

BIPG1198-100MG BIPG1198-250MG
APOSBIPG1198-100MGEA 58 EUR
APOSBIPG1198-100MG APOSBIPG1198-250MG
Biotin-PEG3-acid
Biotin-PEG3-acid
Formel: C₁₉H₃₃N₃O₇S
Molecular Weight: 447.546 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22574817
CAS-Nummer: 252881-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR