Hydroxy-PEG3-acid

Lieferant: Apollo Scientific

Synonyme: 3-(2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy)propanoic acid

BIPG1429-250MG BIPG1429-1G
APOSBIPG1429-250MGEA 82.8 EUR
APOSBIPG1429-250MG APOSBIPG1429-1G
Hydroxy-PEG3-acid
Hydroxy-PEG3-acid
Formel: C₉H₁₈O₆
Molecular Weight: 222,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26793743
CAS-Nummer: 518044-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR