Hydroxy-PEG3-CH2CO2tBu

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2-(2-(2-(2-hydroxyethoxy)ethoxy)ethoxy)acetate, Hydroxy-PEG3-CH2-Boc

BIPG1437-1G BIPG1437-500MG BIPG1437-5G
APOSBIPG1437-1GEA 261 EUR
APOSBIPG1437-1G APOSBIPG1437-500MG APOSBIPG1437-5G
Hydroxy-PEG3-CH2CO2tBu
Hydroxy-PEG3-CH2CO2tBu
Formel: C₁₂H₂₄O₆
Molecular Weight: 264,315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27977510
CAS-Nummer: 518044-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR