Hydroxy-PEG8-acid

Lieferant: Apollo Scientific

BIPG1453-250MG BIPG1453-100MG
APOSBIPG1453-250MGEA 264 EUR
APOSBIPG1453-250MG APOSBIPG1453-100MG
Hydroxy-PEG8-acid
Hydroxy-PEG8-acid
Formel: C₁₉H₃₈O₁₁
Molecular Weight: 442.498 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11041105
CAS-Nummer: 937188-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR