Sulbactam sodium
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Sodium (2S-cis)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4,4-dioxide
BIS0150-10G
BIS0150-1G
BIS0150-5G
APOSBIS0150-10GEA
386
EUR
APOSBIS0150-10G
APOSBIS0150-1G
APOSBIS0150-5G
Sulbactam sodium
Sulbactam sodium
Formel:
C₈H₁₀NO₅S·Na Molecular Weight: 255.223 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01750374 CAS-Nummer: 69388-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...