5-Acetylamino-6-amino-3-methyluracil-[13C4,15N3] (AAMU)

Lieferant: Apollo Scientific
Achtung

BISC1001-1MG BISC1001-5MG
APOSBISC1001-1MGEA 0 EUR
APOSBISC1001-1MG APOSBISC1001-5MG
5-Acetylamino-6-amino-3-methyluracil-[13C4,15N3] (AAMU)
5-Acetylamino-6-amino-3-methyluracil-[13C4,15N3] (AAMU)
Formel: [₁₃C]₄C₃H₁₀[₁₅N]₃NO₃
Molecular Weight: 205,13 g/mol
Schmelzpunkt: 305 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11656111
CAS-Nummer: 1173022-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR