Pyridoxamine-[2H3] dihydrochloride

Lieferant: Apollo Scientific
Achtung

BISC1046-1MG BISC1046-5MG
APOSBISC1046-1MGEA 725 EUR
APOSBISC1046-1MG APOSBISC1046-5MG
Pyridoxamine-[2H3] dihydrochloride
Pyridoxamine-[2H3] dihydrochloride
Formel: C₈H₉D₃N₂O₂·2ClH
Molecular Weight: 244,133 g/mol
Schmelzpunkt: 224…226 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD11656105
CAS-Nummer: 1173023-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR