Pyridoxine-[2H3].HCl

Lieferant: Apollo Scientific
Achtung

BISC1047-1MG BISC1047-5MG
APOSBISC1047-1MGEA 571 EUR
APOSBISC1047-1MG APOSBISC1047-5MG
Pyridoxine-[2H3].HCl
Pyridoxine-[2H3].HCl
Formel: C₈H₈D₃NO₃·ClH
Molecular Weight: 208,657 g/mol
Schmelzpunkt: 214 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11656714
EINECS: 200-386-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR