Resorufin-[2H6]
Lieferant: Apollo Scientific
|
Achtung
|
BISC1048-10MG
BISC1048-50MG
APOSBISC1048-10MGEA
604
EUR
APOSBISC1048-10MG
APOSBISC1048-50MG
Resorufin-[2H6]
Resorufin-[2H6]
Formel:
C₁₂HD₆NO₃ Molecular Weight: 219,226 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD11656671 CAS-Nummer: 1196157-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...