Spermine-[2H8].4HCl

Lieferant: Apollo Scientific
Achtung

BISC1051-10MG BISC1051-5MG
APOSBISC1051-10MGEA 0 EUR
APOSBISC1051-10MG APOSBISC1051-5MG
Spermine-[2H8].4HCl
Spermine-[2H8].4HCl
Formel: C₁₀H₁₈D₈N₄·4ClH
Molecular Weight: 356,233 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11656106
CAS-Nummer: 1173022-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR