Ticarcillin disodium salt/Potassium clavulanate mixture (15:1)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Timentin
BIT0152-1G
BIT0152-5G
BIT0152-10G
APOSBIT0152-1GEA
36.4
EUR
APOSBIT0152-1G
APOSBIT0152-5G
APOSBIT0152-10G
Ticarcillin disodium salt/Potassium clavulanate mixture (15:1)
Ticarcillin disodium salt/Potassium clavulanate mixture (15:1)
Inhibitor of bacterial cell wall synthesis. It inhibitsthe crosslinking of peptidoglycan by binding and inactivating of transpeptidases.
Cell culture tested 1500/100mg