(+)-(S)-ar-Turmerone
Lieferant: Apollo Scientific
|
Achtung
|
BIT1044-10MG
BIT1044-25MG
BIT1044-5MG
APOSBIT1044-10MGEA
406
EUR
APOSBIT1044-10MG
APOSBIT1044-25MG
APOSBIT1044-5MG
(+)-(S)-ar-Turmerone
(+)-(S)-ar-Turmerone
Formel:
C₁₅H₂₀O Lagertemperatur: Gefrierschrank |
MDL:
MFCD09031381 CAS-Nummer: 532-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...