Thulium(III) bromide hydrate 99,9%

Lieferant: Apollo Scientific

IN3592-10G IN3592-1G
APOSIN3592-10GEA 328 EUR
APOSIN3592-10G APOSIN3592-1G
Thulium(III) bromide hydrate 99,9%
Thulium(III) bromide hydrate
Formel: Tm·Br₃·H₂O
Schmelzpunkt: 952 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150441
CAS-Nummer: 226419-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR