Yttrium(III) acetate hydrate 99,9%

Lieferant: Apollo Scientific

Synonyme: Yttrium triacetate hydrate

IN3829-50G IN3829-1KG
APOSIN3829-50GEA 128 EUR
APOSIN3829-50G APOSIN3829-1KG
Yttrium(III) acetate hydrate 99,9%
Yttrium(III) acetate hydrate
Formel: (C₂H₃O₂)₃Y·xH₂O
Molecular Weight: 266,04 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150128
CAS-Nummer: 304675-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR