8-Hydrazinoquinoline dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (Quinolin-8-yl)hydrazine dihydrochloride, 8-Hydrazino-1-azanaphthalene dihydrochloride

OR0048-1G OR0048-500MG
APOSOR0048-1GEA 125 EUR
APOSOR0048-1G APOSOR0048-500MG
8-Hydrazinoquinoline dihydrochloride
8-Hydrazinoquinoline dihydrochloride
Formel: C₉H₉N₃·2ClH
Molecular Weight: 232.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00491379
CAS-Nummer: 91004-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR