Phenazine-1-carboxylic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Carboxyphenazine, 1-Carboxydibenzo[b,e]pyrazine

OR01490-100MG OR01490-1G OR01490-250MG
APOSOR01490-100MGEA 116 EUR
APOSOR01490-100MG APOSOR01490-1G APOSOR01490-250MG
Phenazine-1-carboxylic acid
Phenazine-1-carboxylic acid
Formel: C₁₃H₈N₂O₂
Molecular Weight: 224.215 g/mol
Schmelzpunkt: 239…245 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00185184
CAS-Nummer: 2538-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR