Phenyl(phenylamino)acetic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Anilino(phenyl)acetic acid, Phenyl(phenylamino)ethanoic acid

OR01496-1G OR01496-250MG
APOSOR01496-1GEA 307 EUR
APOSOR01496-1G APOSOR01496-250MG
Phenyl(phenylamino)acetic acid 95%
Phenyl(phenylamino)acetic acid
Formel: C₁₄H₁₃NO₂
Molecular Weight: 227,259 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01724775
CAS-Nummer: 3684-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR