2-(Pyridin-4-yl)-1H-indole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(1H-Indol-2-yl)pyridine
OR01566-250MG
OR01566-1G
APOSOR01566-250MGEA
197
EUR
APOSOR01566-250MG
APOSOR01566-1G
2-(Pyridin-4-yl)-1H-indole 95%
2-(Pyridin-4-yl)-1H-indole
Formel:
C₁₃H₁₀N₂ Molecular Weight: 194,232 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04966830 CAS-Nummer: 21182-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...