3-Chloro-4-hydroxytetrahydrothiophene 1,1-dioxide 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chlorotetrahydrothiophene-3-ol 1,1-dioxide, 4-Chloro-3-hydroxythiolane 1,1-dioxide
OR015663-1G
OR015663-500MG
APOSOR015663-1GEA
618
EUR
APOSOR015663-1G
APOSOR015663-500MG
3-Chloro-4-hydroxytetrahydrothiophene 1,1-dioxide 95%
3-Chloro-4-hydroxytetrahydrothiophene 1,1-dioxide
Formel:
C₄H₇ClO₃S Molecular Weight: 170,615 g/mol Schmelzpunkt: 163…167 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005486 CAS-Nummer: 49592-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...