3-(1-Hydroxyethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Pyridin-3-yl)ethan-1-ol, 1-Hydroxy-1-(pyridin-3-yl)ethane
OR01622-25G
OR01622-5G
APOSOR01622-25GEA
287
EUR
APOSOR01622-25G
APOSOR01622-5G
3-(1-Hydroxyethyl)pyridine
3-(1-Hydroxyethyl)pyridine
Formel:
C₇H₉NO Molecular Weight: 123,152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01691686 CAS-Nummer: 4754-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...