6-[2-(Pyridin-3-yl)pyrrolidin-1-yl]nicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(5-Carboxypyridin-2-yl)-2-(pyridin-3-yl)pyrrolidine, 6-[2-(Pyridin-3-yl)pyrrolidin-1-yl]pyridine-3-carboxylic acid

OR01741-10MG OR01741-1G
APOSOR01741-10MGEA 36.4 EUR
APOSOR01741-10MG APOSOR01741-1G
6-[2-(Pyridin-3-yl)pyrrolidin-1-yl]nicotinic acid
6-[2-(Pyridin-3-yl)pyrrolidin-1-yl]nicotinic acid
Formel: C₁₅H₁₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06245472
CAS-Nummer: 904817-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR