3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR018945-1G OR018945-250MG
APOSOR018945-1GEA 0 EUR
APOSOR018945-1G APOSOR018945-250MG
3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid 97%
3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid
Formel: C₁₃H₈ClNO₂S₂
Schmelzpunkt: 241…243 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00112478
CAS-Nummer: 116525-66-7
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR