6-Amino-2,3-dichlorobenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Cyano-3,4-dichloroaniline
OR01914-100G
OR01914-25G
OR01914-5G
APOSOR01914-100GEA
1330
EUR
APOSOR01914-100G
APOSOR01914-25G
APOSOR01914-5G
6-Amino-2,3-dichlorobenzonitrile
6-Amino-2,3-dichlorobenzonitrile
Formel:
C₇H₄Cl₂N₂ Schmelzpunkt: 141…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00728602 CAS-Nummer: 147249-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...