3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Carboxy-3-chloroimidazo[1,2-a]pyridine
OR01957-1G
OR01957-250MG
APOSOR01957-1GEA
157
EUR
APOSOR01957-1G
APOSOR01957-250MG
3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel:
C₈H₅ClN₂O₂ Molecular Weight: 196.591 g/mol Schmelzpunkt: 198.5…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09836169 CAS-Nummer: 1000017-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...