3-Methoxybenzylhydrazine dihydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Hydrazinobenzyl methyl ether dihydrochloride, (3-Methoxybenzyl)hydrazine dihydrochloride, 3-(Hydrazinomethyl)phenyl methyl ether dihydrochloride, 3-(Hydrazinomethyl)anisole dihydrochloride

OR0202-1G OR0202-5G OR0202-25G
APOSOR0202-1GEA 208 EUR
APOSOR0202-1G APOSOR0202-5G APOSOR0202-25G
3-Methoxybenzylhydrazine dihydrochloride 95+%
3-Methoxybenzylhydrazine dihydrochloride
Formel: C₈H₁₂N₂O·2ClH
Molecular Weight: 225.116 g/mol
Schmelzpunkt: 115…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06245507
CAS-Nummer: 849021-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR