2-(Piperazin-1-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Carboxyphenyl)piperazine

OR0215-10G OR0215-1G OR0215-5G
APOSOR0215-10GEA 244 EUR
APOSOR0215-10G APOSOR0215-1G APOSOR0215-5G
2-(Piperazin-1-yl)benzoic acid 97%
2-(Piperazin-1-yl)benzoic acid
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206,241 g/mol
Schmelzpunkt: 243 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095640
CAS-Nummer: 446831-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR