6-Chloro-3-phenyl[1,2,4]triazolo[4,3-b]pyridazine

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3-yl)benzene

OR0294-1G OR0294-250MG
APOSOR0294-1GEA 306 EUR
APOSOR0294-1G APOSOR0294-250MG
6-Chloro-3-phenyl[1,2,4]triazolo[4,3-b]pyridazine
6-Chloro-3-phenyl[1,2,4]triazolo[4,3-b]pyridazine
Formel: C₁₁H₇ClN₄
Molecular Weight: 230,653 g/mol
Schmelzpunkt: 201…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00277891
CAS-Nummer: 7190-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR