3-(Cyclopentyloxy)-4-methoxybenzaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Cyclopentyloxy)-4-formylanisole

OR0323-50MG OR0323-100MG
APOSOR0323-50MGEA 21.9 EUR
APOSOR0323-50MG APOSOR0323-100MG
3-(Cyclopentyloxy)-4-methoxybenzaldehyde
3-(Cyclopentyloxy)-4-methoxybenzaldehyde
Formel: C₁₃H₁₆O₃
Molecular Weight: 220,264 g/mol
Siedepunkt: 124…126 °C (0,05 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00673136
CAS-Nummer: 67387-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR