Indane-2-isothiocyanate

Lieferant: Apollo Scientific

OR0546-1G OR0546-250MG OR0546-500MG
APOSOR0546-1GEA 1040 EUR
APOSOR0546-1G APOSOR0546-250MG APOSOR0546-500MG
Indane-2-isothiocyanate
Indane-2-isothiocyanate
Formel: C₁₀H₉NS
Molecular Weight: 175,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01313225
CAS-Nummer: 306935-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR