Methyl adamant-1-ylacetate
Lieferant: Apollo Scientific
|
Achtung
|
OR0596-10G
OR0596-1G
OR0596-5G
APOSOR0596-10GEA
346
EUR
APOSOR0596-10G
APOSOR0596-1G
APOSOR0596-5G
Methyl adamant-1-ylacetate
Methyl adamant-1-ylacetate
Formel:
C₁₃H₂₀O₂ Molecular Weight: 208,297 g/mol Siedepunkt: 86 °C (0,3 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01073581 CAS-Nummer: 27174-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...