Methyl 2-(4-isobutylphenyl)propanoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-[4-(2-methylprop-1-yl)phenyl]propionate, Methyl2-(4-isobutylphenyl)propanoate, Methyl 2-[4-(2-methylprop-1-yl)phenyl]propanoate
OR0631-1G
OR0631-250MG
APOSOR0631-1GEA
29
EUR
APOSOR0631-1G
APOSOR0631-250MG
Methyl 2-(4-isobutylphenyl)propanoate
Methyl 2-(4-isobutylphenyl)propanoate
Formel:
C₁₄H₂₀O₂ Molecular Weight: 220,307 g/mol Siedepunkt: 100 °C (0,8 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00869922 CAS-Nummer: 61566-34-5 EINECS: 262-853-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...