3-Aminopiperidine hydrochloride, N1-CBZ protected

Lieferant: Apollo Scientific

Synonyme: Benzyl 3-aminopiperidine-1-carboxylate hydrochloride, 3-Amino-1-benzyloxycarbonylpiperidine hydrochloride

OR0734-1G OR0734-5G
APOSOR0734-1GEA 82.8 EUR
APOSOR0734-1G APOSOR0734-5G
3-Aminopiperidine hydrochloride, N1-CBZ protected
3-Aminopiperidine hydrochloride, N1-CBZ protected
Formel: C₁₃H₁₈N₂O₂·ClH
Molecular Weight: 270,755 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038445
CAS-Nummer: 960541-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR