3-Aminopiperidine hydrochloride, N1-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-aminopiperidine-1-carboxylate hydrochloride, 3-Amino-1-(tert-butoxycarbonyl)piperidine hydrochloride

OR0735-10G OR0735-1G OR0735-5G
APOSOR0735-10GEA 487 EUR
APOSOR0735-10G APOSOR0735-1G APOSOR0735-5G
3-Aminopiperidine hydrochloride, N1-BOC protected
3-Aminopiperidine hydrochloride, N1-BOC protected
Formel: C₁₀H₂₀N₂O₂·ClH
Molecular Weight: 236,739 g/mol
Schmelzpunkt: 181…182 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD06691424
CAS-Nummer: 1000796-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR