Tetramethylsuccinic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,2,3,3-Tetramethylbutane-1,4-dioic acid

OR0747-1G OR0747-250MG
APOSOR0747-1GEA 406 EUR
APOSOR0747-1G APOSOR0747-250MG
Tetramethylsuccinic acid
Tetramethylsuccinic acid
Formel: C₈H₁₄O₄
Molecular Weight: 174,194 g/mol
Schmelzpunkt: 204…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00829122
CAS-Nummer: 630-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR