4-(Benzylamino)piperidine, N1-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-(benzylamino)piperidine-1-carboxylate, 1-Boc-4-Benzylaminopiperidine, 4-(Benzylamino)-1-(tert-butoxycarbonyl)piperidine

OR0810-1G OR0810-5G
APOSOR0810-1GEA 105 EUR
APOSOR0810-1G APOSOR0810-5G
4-(Benzylamino)piperidine, N1-BOC protected
4-(Benzylamino)piperidine, N1-BOC protected
Formel: C₁₇H₂₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04114987
CAS-Nummer: 206273-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR