N,N'-Dimethyldodecane-1,12-diamine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR0811-10G
OR0811-2G
APOSOR0811-10GEA
348
EUR
APOSOR0811-10G
APOSOR0811-2G
N,N'-Dimethyldodecane-1,12-diamine 97%
N,N'-Dimethyldodecane-1,12-diamine
Formel:
C₁₄H₃₂N₂ Siedepunkt: 118…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095638 CAS-Nummer: 56992-91-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...