1-(2,2-Dimethylpropanoyl)piperazine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2-Dimethyl-1-(piperazin-1-yl)propan-1-one, 1-Pivaloylpiperazine
OR0851-1G
OR0851-5G
APOSOR0851-1GEA
151
EUR
APOSOR0851-1G
APOSOR0851-5G
1-(2,2-Dimethylpropanoyl)piperazine
1-(2,2-Dimethylpropanoyl)piperazine
Formel:
C₉H₁₈N₂O Molecular Weight: 170,252 g/mol Schmelzpunkt: 55…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973343 CAS-Nummer: 155295-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...