1-(Piperazin-1-yl)propan-1-one 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Propanoylpiperazine, 1-(1-Oxoprop-1-yl)piperazine

OR0854-1G OR0854-250MG
APOSOR0854-1GEA 31.9 EUR
APOSOR0854-1G APOSOR0854-250MG
1-(Piperazin-1-yl)propan-1-one 98%
1-(Piperazin-1-yl)propan-1-one
Formel: C₇H₁₄N₂O
Molecular Weight: 142,199 g/mol
Siedepunkt: 78 °C (0,2 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD04973346
CAS-Nummer: 76816-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR