2-Methyl-1-(piperazin-1-yl)propan-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Methylpropionoyl)piperazine, 1-Isobutanoylpiperazine, 1-(2-Methylpropanoyl)piperazine

OR0858-5G
APOSOR0858-5GEA 48 EUR
APOSOR0858-5G
2-Methyl-1-(piperazin-1-yl)propan-1-one
2-Methyl-1-(piperazin-1-yl)propan-1-one
Formel: C₈H₁₆N₂O
Molecular Weight: 156,225 g/mol
Siedepunkt: 76…78 °C (0,15 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD04973344
CAS-Nummer: 71260-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR