1-(Biphenyl-3-yl)piperazine

Lieferant: Apollo Scientific
Achtung

OR0864-1G OR0864-5G
APOSOR0864-1GEA 226 EUR
APOSOR0864-1G APOSOR0864-5G
1-(Biphenyl-3-yl)piperazine
1-(Biphenyl-3-yl)piperazine
Formel: C₁₆H₁₈N₂
Molecular Weight: 238.328 g/mol
Schmelzpunkt: 69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06201887
CAS-Nummer: 115761-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR