1-(3-Phenoxypropyl)piperazine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[3-(Piperazin-1-yl)propoxy]benzene
OR0892-1G
OR0892-10G
OR0892-5G
APOSOR0892-1GEA
114
EUR
APOSOR0892-1G
APOSOR0892-10G
APOSOR0892-5G
1-(3-Phenoxypropyl)piperazine 98%
1-(3-Phenoxypropyl)piperazine
Formel:
C₁₃H₂₀N₂O Molecular Weight: 220,311 g/mol Schmelzpunkt: 48…51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844730 CAS-Nummer: 41298-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...