1-(2-Propoxyethyl)piperazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-{[2-(Prop-1-yl)oxy]ethyl}piperazine

OR0955-250MG OR0955-2G
APOSOR0955-250MGEA 43.5 EUR
APOSOR0955-250MG APOSOR0955-2G
1-(2-Propoxyethyl)piperazine
1-(2-Propoxyethyl)piperazine
Formel: C₉H₂₀N₂O
Molecular Weight: 172,268 g/mol
Siedepunkt: 107 °C (12 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD04117823
CAS-Nummer: 741667-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR